Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 71908]

Supplier: R & D Systems
MAB6311
220021-373EA 763.57 USD
220021-373
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 71908]
Antibodies
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 71908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR