Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 71903]

Supplier: R & D Systems
MAB631-500
MSPP-MAB631500EA 1459.17 USD
MSPP-MAB631500
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 71903]
Antibodies
Agonist Activity

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 71903
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR