Anti-COL4A1 Mouse Monoclonal Antibody [clone: 577238]

Supplier: R & D Systems
MAB6308
102886-344EA 633.42 USD
102886-344
Anti-COL4A1 Mouse Monoclonal Antibody [clone: 577238]
Antibodies
Type: Primary
Antigen: COL4A1
Clonality: Monoclonal
Clone: 577238
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR