Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 90906]

Supplier: R & D Systems
MAB6302
220021-371EA 763.57 USD
220021-371
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 90906]
Antibodies
Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: 90906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR