Anti-MUC1 Mouse Monoclonal Antibody [clone: 604804]

Supplier: R & D Systems
MAB6298
102886-340EA 655.11 USD
102886-340
Anti-MUC1 Mouse Monoclonal Antibody [clone: 604804]
Antibodies
Simple Western

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: 604804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR