Anti-PPY Mouse Monoclonal Antibody [clone: 548301]

Supplier: R & D Systems
MAB6297
102886-336EA 566.89 USD
102886-336
Anti-PPY Mouse Monoclonal Antibody [clone: 548301]
Antibodies
Type: Primary
Antigen: PPY
Clonality: Monoclonal
Clone: 548301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR