Anti-TDRD1 Rat Monoclonal Antibody [clone: 739206]

Supplier: R & D Systems
MAB6296
102886-334EA 577.02 USD
102886-334
Anti-TDRD1 Rat Monoclonal Antibody [clone: 739206]
Antibodies
Type: Primary
Antigen: TDRD1
Clonality: Monoclonal
Clone: 739206
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR