Anti-ONECUT2 Mouse Monoclonal Antibody [clone: 606014]

Supplier: R & D Systems
MAB6294
102886-332EA 594.37 USD
102886-332
Anti-ONECUT2 Mouse Monoclonal Antibody [clone: 606014]
Antibodies
Type: Primary
Antigen: ONECUT2
Clonality: Monoclonal
Clone: 606014
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR