Anti-LTBR Mouse Monoclonal Antibody [clone: 71319]

Supplier: R & D Systems
MAB629
220021-368EA 772.25 USD
220021-368
Anti-LTBR Mouse Monoclonal Antibody [clone: 71319]
Antibodies
Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 71319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR