Anti-DFFB Mouse Monoclonal Antibody [clone: 609809]

Supplier: R & D Systems
MAB6289
220021-367EA 561.11 USD
220021-367
Anti-DFFB Mouse Monoclonal Antibody [clone: 609809]
Antibodies
Type: Primary
Antigen: DFFB
Clonality: Monoclonal
Clone: 609809
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR