Anti-LILRA4 Mouse Monoclonal Antibody [clone: 656688]

Supplier: R & D Systems
MAB6287
102886-330EA 600.15 USD
102886-330
Anti-LILRA4 Mouse Monoclonal Antibody [clone: 656688]
Antibodies
Type: Primary
Antigen: LILRA4
Clonality: Monoclonal
Clone: 656688
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR