Anti-TDRKH Mouse Monoclonal Antibody [clone: 686213]

Supplier: R & D Systems
MAB6286
102886-328EA 577.02 USD
102886-328
Anti-TDRKH Mouse Monoclonal Antibody [clone: 686213]
Antibodies
Type: Primary
Antigen: TDRKH
Clonality: Monoclonal
Clone: 686213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR