Anti-JADE1 Mouse Monoclonal Antibody [clone: 658005]

Supplier: R & D Systems
MAB6275
102886-324EA 594.37 USD
102886-324
Anti-JADE1 Mouse Monoclonal Antibody [clone: 658005]
Antibodies
Type: Primary
Antigen: JADE1
Clonality: Monoclonal
Clone: 658005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR