Anti-WFDC2 Mouse Monoclonal Antibody [clone: 676013]

Supplier: R & D Systems
MAB6274
102886-322EA 633.42 USD
102886-322
Anti-WFDC2 Mouse Monoclonal Antibody [clone: 676013]
Antibodies
Type: Primary
Antigen: WFDC2
Clonality: Monoclonal
Clone: 676013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR