Anti-CD6 Mouse Monoclonal Antibody [clone: 123119]

Supplier: R & D Systems
MAB627
220021-365EA 772.25 USD
220021-365
Anti-CD6 Mouse Monoclonal Antibody [clone: 123119]
Antibodies
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 123119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR