Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 685809]

Supplier: R & D Systems
MAB6263
102886-320EA 594.37 USD
102886-320
Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 685809]
Antibodies
Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 685809
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR