Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 175707]

Supplier: R & D Systems
MAB6262
220021-364EA 772.25 USD
220021-364
Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 175707]
Antibodies
Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 175707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR