Anti-LIMS1 Mouse Monoclonal Antibody [clone: 591604]

Supplier: R & D Systems
MAB6257
220021-361EA 561.11 USD
220021-361
Anti-LIMS1 Mouse Monoclonal Antibody [clone: 591604]
Antibodies
Type: Primary
Antigen: LIMS1
Clonality: Monoclonal
Clone: 591604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR