Anti-NME1 Mouse Monoclonal Antibody [clone: 642817]

Supplier: R & D Systems
MAB6256
220021-360EA 577.02 USD
220021-360
Anti-NME1 Mouse Monoclonal Antibody [clone: 642817]
Antibodies
Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 642817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR