Anti-TLR4 Mouse Monoclonal Antibody [clone: 610015]

Supplier: R & D Systems
MAB6248
102886-318EA 594.37 USD
102886-318
Anti-TLR4 Mouse Monoclonal Antibody [clone: 610015]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 610015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR