Anti-ACPP Mouse Monoclonal Antibody [clone: 6917]

Supplier: R & D Systems
MAB6240
102886-312EA 633.42 USD
102886-312
Anti-ACPP Mouse Monoclonal Antibody [clone: 6917]
Antibodies
Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: 6917
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR