Anti-HEXA Mouse Monoclonal Antibody [clone: 714712]

Supplier: R & D Systems
MAB62371
102886-310EA 577.02 USD
102886-310
Anti-HEXA Mouse Monoclonal Antibody [clone: 714712]
Antibodies
Type: Primary
Antigen: HEXA
Clonality: Monoclonal
Clone: 714712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR