Anti-LRRC32 Rat Monoclonal Antibody [clone: 725226]

Supplier: R & D Systems
MAB62291
102886-306EA 594.37 USD
102886-306
Anti-LRRC32 Rat Monoclonal Antibody [clone: 725226]
Antibodies
Type: Primary
Antigen: LRRC32
Clonality: Monoclonal
Clone: 725226
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR