Anti-LAMP2 Mouse Monoclonal Antibody [clone: 743320]

Supplier: R & D Systems
MAB6228
102886-304EA 594.37 USD
102886-304
Anti-LAMP2 Mouse Monoclonal Antibody [clone: 743320]
Antibodies
Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: 743320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR