Anti-TRIM21 Mouse Monoclonal Antibody [clone: 672714]

Supplier: R & D Systems
MAB62191
102886-302EA 594.37 USD
102886-302
Anti-TRIM21 Mouse Monoclonal Antibody [clone: 672714]
Antibodies
Type: Primary
Antigen: TRIM21
Clonality: Monoclonal
Clone: 672714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR