Anti-TICAM1 Mouse Monoclonal Antibody [clone: 567212]

Supplier: R & D Systems
MAB6216
102886-296EA 594.37 USD
102886-296
Anti-TICAM1 Mouse Monoclonal Antibody [clone: 567212]
Antibodies
Type: Primary
Antigen: TICAM1
Clonality: Monoclonal
Clone: 567212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR