Anti-CASP1 Mouse Monoclonal Antibody [clone: 661228]

Supplier: R & D Systems
MAB6215
102886-294EA 633.42 USD
102886-294
Anti-CASP1 Mouse Monoclonal Antibody [clone: 661228]
Antibodies
Type: Primary
Antigen: CASP1
Clonality: Monoclonal
Clone: 661228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR