Anti-PTK2B Mouse Monoclonal Antibody [clone: 592918]

Supplier: R & D Systems
MAB6210
220021-356EA 594.37 USD
220021-356
Anti-PTK2B Mouse Monoclonal Antibody [clone: 592918]
Antibodies
Simple Western

Type: Primary
Antigen: PTK2B
Clonality: Monoclonal
Clone: 592918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR