Anti-KPNA2 Mouse Monoclonal Antibody [clone: 682208]

Supplier: R & D Systems
MAB62071
102886-292EA 561.11 USD
102886-292
Anti-KPNA2 Mouse Monoclonal Antibody [clone: 682208]
Antibodies
Type: Primary
Antigen: KPNA2
Clonality: Monoclonal
Clone: 682208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR