Anti-KPNA2 Mouse Monoclonal Antibody [clone: 682239]

Supplier: R & D Systems
MAB6207
102886-290EA 600.15 USD
102886-290
Anti-KPNA2 Mouse Monoclonal Antibody [clone: 682239]
Antibodies
Type: Primary
Antigen: KPNA2
Clonality: Monoclonal
Clone: 682239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR