Anti-CMV UL146/vCXC1 Mouse Monoclonal Antibody [clone: 76520]

Supplier: R & D Systems
MAB620
220021-354EA 772.25 USD
220021-354
Anti-CMV UL146/vCXC1 Mouse Monoclonal Antibody [clone: 76520]
Antibodies
Type: Primary
Antigen: CMV UL146/vCXC1
Clonality: Monoclonal
Clone: 76520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR