Anti-CD96 Mouse Monoclonal Antibody [clone: 628226]

Supplier: R & D Systems
MAB61991
102886-286EA 577.02 USD
102886-286
Anti-CD96 Mouse Monoclonal Antibody [clone: 628226]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 628226
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR