Anti-CD96 Mouse Monoclonal Antibody [clone: 628211]

Supplier: R & D Systems
MAB6199
102886-284EA 647.88 USD
102886-284
Anti-CD96 Mouse Monoclonal Antibody [clone: 628211]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 628211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR