Anti-HAO1 Mouse Monoclonal Antibody [clone: 732410]

Supplier: R & D Systems
MAB6197
102886-280EA 594.37 USD
102886-280
Anti-HAO1 Mouse Monoclonal Antibody [clone: 732410]
Antibodies
Type: Primary
Antigen Symbol: HAO1
Antigen Name: hydroxyacid oxidase (glycolate oxidase) 1
Antigen Synonyms: GOX, GOX1, HAOX1
Clonality: Monoclonal
Clone: 732410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR