Anti-ITGA8 Mouse Monoclonal Antibody [clone: 481709]

Supplier: R & D Systems
MAB6194
102886-276EA 594.37 USD
102886-276
Anti-ITGA8 Mouse Monoclonal Antibody [clone: 481709]
Antibodies
Type: Primary
Antigen: ITGA8
Clonality: Monoclonal
Clone: 481709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR