Anti-TIE1 Mouse Monoclonal Antibody [clone: 88016]

Supplier: R & D Systems
MAB619
220021-353EA 772.25 USD
220021-353
Anti-TIE1 Mouse Monoclonal Antibody [clone: 88016]
Antibodies
Type: Primary
Antigen: TIE1
Clonality: Monoclonal
Clone: 88016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR