Anti-VWA1 Mouse Monoclonal Antibody [clone: 559915]

Supplier: R & D Systems
MAB6189
220021-352EA 561.11 USD
220021-352
Anti-VWA1 Mouse Monoclonal Antibody [clone: 559915]
Antibodies
Type: Primary
Antigen: VWA1
Clonality: Monoclonal
Clone: 559915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR