Anti-UNC13B Mouse Monoclonal Antibody [clone: 584102]

Supplier: R & D Systems
MAB6188
220021-351EA 577.02 USD
220021-351
Anti-UNC13B Mouse Monoclonal Antibody [clone: 584102]
Antibodies
Type: Primary
Antigen: UNC13B
Clonality: Monoclonal
Clone: 584102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR