Anti-BMP5 Rat Monoclonal Antibody [clone: 781321]

Supplier: R & D Systems
MAB6176
102886-272EA 577.02 USD
102886-272
Anti-BMP5 Rat Monoclonal Antibody [clone: 781321]
Antibodies
Type: Primary
Antigen: BMP5
Clonality: Monoclonal
Clone: 781321
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR