Anti-PRDM14 Mouse Monoclonal Antibody [clone: 647002]

Supplier: R & D Systems
MAB6175
102886-270EA 577.02 USD
102886-270
Anti-PRDM14 Mouse Monoclonal Antibody [clone: 647002]
Antibodies
Type: Primary
Antigen: PRDM14
Clonality: Monoclonal
Clone: 647002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR