Anti-SULT2B1 Mouse Monoclonal Antibody [clone: 682516]

Supplier: R & D Systems
MAB6174
102886-268EA 561.11 USD
102886-268
Anti-SULT2B1 Mouse Monoclonal Antibody [clone: 682516]
Antibodies
Type: Primary
Antigen: SULT2B1
Clonality: Monoclonal
Clone: 682516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR