Anti-EOMES Mouse Monoclonal Antibody [clone: 644730]

Supplier: R & D Systems
MAB6166
102886-266EA 655.11 USD
102886-266
Anti-EOMES Mouse Monoclonal Antibody [clone: 644730]
Antibodies
Type: Primary
Antigen: EOMES
Clonality: Monoclonal
Clone: 644730
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR