Anti-FOXO3 Mouse Monoclonal Antibody [clone: 648716]

Supplier: R & D Systems
MAB6165
102886-264EA 633.42 USD
102886-264
Anti-FOXO3 Mouse Monoclonal Antibody [clone: 648716]
Antibodies
Type: Primary
Antigen Symbol: FOXO3
Antigen Name: forkhead box O3
Antigen Synonyms: 1110048B16Rik, 2010203A17Rik, C76856, FKHRL1, Fkhr2, Foxo3a
Clonality: Monoclonal
Clone: 648716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR