Anti-PXN Mouse Monoclonal Antibody [clone: 698239]

Supplier: R & D Systems
MAB61641
102886-262EA 566.89 USD
102886-262
Anti-PXN Mouse Monoclonal Antibody [clone: 698239]
Antibodies
Type: Primary
Antigen: PXN
Clonality: Monoclonal
Clone: 698239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR