Anti-PXN Mouse Monoclonal Antibody [clone: 601327]

Supplier: R & D Systems
MAB6164
220021-348EA 594.37 USD
220021-348
Anti-PXN Mouse Monoclonal Antibody [clone: 601327]
Antibodies
Type: Primary
Antigen: PXN
Clonality: Monoclonal
Clone: 601327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR