Anti-UCP1 Mouse Monoclonal Antibody [clone: 536435]

Supplier: R & D Systems
MAB6158
220021-346EA 655.11 USD
220021-346
Anti-UCP1 Mouse Monoclonal Antibody [clone: 536435]
Antibodies
Simple Western

Type: Primary
Antigen: UCP1
Clonality: Monoclonal
Clone: 536435
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR