Anti-CST3 Mouse Monoclonal Antibody [clone: 678920]

Supplier: R & D Systems
MAB6154
102886-260EA 561.11 USD
102886-260
Anti-CST3 Mouse Monoclonal Antibody [clone: 678920]
Antibodies
Type: Primary
Antigen: CST3
Clonality: Monoclonal
Clone: 678920
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR