Anti-PRTN3 Mouse Monoclonal Antibody [clone: 684042]

Supplier: R & D Systems
MAB6134
102886-250EA 594.37 USD
102886-250
Anti-PRTN3 Mouse Monoclonal Antibody [clone: 684042]
Antibodies
Type: Primary
Antigen: PRTN3
Clonality: Monoclonal
Clone: 684042
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR