Anti-SEMA4C Mouse Monoclonal Antibody [clone: 660016]

Supplier: R & D Systems
MAB6125
102886-246EA 594.37 USD
102886-246
Anti-SEMA4C Mouse Monoclonal Antibody [clone: 660016]
Antibodies
Type: Primary
Antigen: SEMA4C
Clonality: Monoclonal
Clone: 660016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR