Anti-IZUMO1R Rat Monoclonal Antibody [clone: 721601]

Supplier: R & D Systems
MAB6124
102886-244EA 561.11 USD
102886-244
Anti-IZUMO1R Rat Monoclonal Antibody [clone: 721601]
Antibodies
Type: Primary
Antigen: IZUMO1R
Clonality: Monoclonal
Clone: 721601
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR